Learn More
Cayman Chemical CA-074 methyl ester 500ug
SDP
Catalog No. 502967687
Encompass_Preferred
A cell-permeable inhibitor of cathepsin B and L in vivo and in whole cells; used to study the role of cathepsin B in neurodegeneration, cancer cell metastasis, necrosis, and rheumatoid arthritis
Catalog No. 50-296-7687 Supplier Cayman Chemical Supplier No. 18469500 UG
A cell-permeable inhibitor of cathepsin B and L in vivo and in whole cells; used to study the role of cathepsin B in neurodegeneration, cancer cell metastasis, necrosis, and rheumatoid arthritis