Learn More
TARGETMOL CHEMICALS INC CA-074 METHYL ESTER 5MG
SDP
Catalog No. 502576946
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CA-074 methyl ester (Cathepsin B Inhibitor IV) is a specific inhibitor of Cathepsin B with neuroprotective anti-cancer and anti-inflammatory effects. purity: 99%
Catalog No. 50-257-6946 Supplier TARGETMOL CHEMICALS INC Supplier No. T34205MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CA-074 methyl ester (Cathepsin B Inhibitor IV) is a specific inhibitor of Cathepsin B with neuroprotective anti-cancer and anti-inflammatory effects. purity: 99%