Learn More
Selleck Chemical LLC CA-074 methyl ester (CA-074 Me) 5mg 147859-80-1 Cathepsin B Inhibitor IV
SDP
Catalog No. 501366348
Encompass_Preferred
CA-074 methyl ester (CA-074 Me, Cathepsin B Inhibitor IV) is a membrane-permeable derivative of CA-074 and acts as an irreversible cathepsin B inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6348 Supplier Selleck Chemical LLC Supplier No. S74205MG
CA-074 methyl ester (CA-074 Me, Cathepsin B Inhibitor IV) is a membrane-permeable derivative of CA-074 and acts as an irreversible cathepsin B inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.