Learn More
Selleck Chemical LLC CA-074 methyl ester
SDP
Catalog No. 5000001418
Encompass_Preferred
CA-074 methyl ester (CA-074 Me Cathepsin B Inhibitor IV) is a membrane-permeable derivative of CA-074 and acts as an irreversible cathepsin B inhibitor
Catalog No. 50-000-01418 Supplier Selleck Chemical LLC Supplier No. S7420100MG
CA-074 methyl ester (CA-074 Me Cathepsin B Inhibitor IV) is a membrane-permeable derivative of CA-074 and acts as an irreversible cathepsin B inhibitor