Learn More
Carbosynth LLC. Mouse monoclonal CA 15-3 antibody
SDP
Supplier:  Carbosynth LLC. 10C03E1MG
Encompass_Preferred
CA 15-3 is breast tumor marker. Diagnostic tests measuring the MUC1- glycoprotein are FDA/EU approved including CA 15-3, CA 27-29 and Br assays. Reoccurrence detection of breast cancer by MUC-1 assay can preceed confirmation by radiological detection methods by 1-9 months.
Catalog No. 50-125-2515