Learn More
Diagnostic Biosystems CA 19-9 CA 19-9
Catalog No. 502407508
Encompass_Preferred
This antibody reacts specifically with sialyl Lewisa and recognizes an epitope being designated CA 19-9. It does not cross-reacts with Lewisa and Lewisb. CA 19-9 has been shown to be useful marker in the diagnosis and management of gastrointestinal cancers.
Catalog No. 50-240-7508 Supplier Diagnostic Biosystems Supplier No. PDM567
This antibody reacts specifically with sialyl Lewisa and recognizes an epitope being designated CA 19-9. It does not cross-reacts with Lewisa and Lewisb. CA 19-9 has been shown to be useful marker in the diagnosis and management of gastrointestinal cancers.