Learn More
Medchemexpress LLC HY-135317 5mg Medchemexpress, CA-4948 CAS:1801344-14-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1353175MG
Encompass_Preferred
Medchemexpress, HY-135317 5mg CA-4948 CAS:1801344-14-8 CA-4948 is a potent IRAK4/FLT3 inhibtor with anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0334