Learn More
Medchemexpress LLC HY-112698 10mg Medchemexpress, CA-5f CAS:1370032-19-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11269810MG
Encompass_Preferred
Medchemexpress, HY-112698 10mg CA-5f CAS:1370032-19-1 CA-5f is a potent late-stage macroautophagy/autophagy inhibitor via inhibiting autophagosome-lysosome fusion. CA-5f increases LC3B-II (a marker to monitor autophagy) and SQSTM1 protein, and also increases ROS production. Anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9144