Learn More
Medchemexpress LLC CA-5f | 1370032-19-1 | 99.1% | 388.46 | 1 ML
SDP
Catalog No. 5000438545
Encompass_Preferred
CA-5f is a potent late-stage macroautophagy/autophagy inhibitor, working by inhibiting autophagosome-lysosome fusion. It increases LC3B-II and SQSTM1 protein levels and also increases ROS production, demonstrating anti-tumor activity.
  • Potent late-stage macroautophagy/autophagy inhibitor
  • Inhibits autophagosome-lysosome fusion
  • Increases LC3B-II protein and SQSTM1 protein
  • Increases ROS production
  • Exhibits anti-tumor activity
  • Inhibits the growth of A549 cells, and is less cytotoxic to normal HUVECs in vitro
  • Does not impair the hydrolytic function or quantity of lysosomes in vitro
  • Well tolerated and potently inhibits tumor growth in nude mice bearing A549 lung cancer cells in vivo
  • Suppresses autophagic flux and induces apoptosis in nude mice bearing A549 lung cancer cells in vivo
Catalog No. 50-004-38545 Supplier Medchemexpress LLC Supplier No. HY11269810MM/1ML
CA-5f is a potent late-stage macroautophagy/autophagy inhibitor, working by inhibiting autophagosome-lysosome fusion. It increases LC3B-II and SQSTM1 protein levels and also increases ROS production, demonstrating anti-tumor activity.
  • Potent late-stage macroautophagy/autophagy inhibitor
  • Inhibits autophagosome-lysosome fusion
  • Increases LC3B-II protein and SQSTM1 protein
  • Increases ROS production
  • Exhibits anti-tumor activity
  • Inhibits the growth of A549 cells, and is less cytotoxic to normal HUVECs in vitro
  • Does not impair the hydrolytic function or quantity of lysosomes in vitro
  • Well tolerated and potently inhibits tumor growth in nude mice bearing A549 lung cancer cells in vivo
  • Suppresses autophagic flux and induces apoptosis in nude mice bearing A549 lung cancer cells in vivo