Learn More
Selleck Chemical LLC CA-5f
SDP
Catalog No. 5000003828
Encompass_Preferred
CA-5f is a potent late-stage macroautophagy/autophagy inhibitor via inhibiting autophagosome-lysosome fusion
Catalog No. 50-000-03828 Supplier Selleck Chemical LLC Supplier No. S592025MG
CA-5f is a potent late-stage macroautophagy/autophagy inhibitor via inhibiting autophagosome-lysosome fusion