Learn More
Biotium Calcium Calibration Buffer
Catalog No. 501964714
Encompass_Preferred
This kit provides calibration buffers and a protocol for producing a range of accurate calcium concentrations for the calibration of fluorescent Ca2 indicators. Each kit contains 50 mL each of components A and B: Component A (zero free Ca2 ): Zero mM CaEGTA (10 mM K2EGTA,100 mM KCI and 10 mM MOPS, pH 7.2) Component B (40 uM free Ca2 ): 10 mM CaEGTA (10 mM CaEGTA,100 mM KCI and 10 mM MOPS, pH 7.2)Properties: Colorless liquids; Store at 4°C; Preservative-free, use within three months of receipt.
Catalog No. 50-196-4714 Supplier Biotium Supplier No. 59100
This kit provides calibration buffers and a protocol for producing a range of accurate calcium concentrations for the calibration of fluorescent Ca2 indicators. Each kit contains 50 mL each of components A and B: Component A (zero free Ca2 ): Zero mM CaEGTA (10 mM K2EGTA,100 mM KCI and 10 mM MOPS, pH 7.2) Component B (40 uM free Ca2 ): 10 mM CaEGTA (10 mM CaEGTA,100 mM KCI and 10 mM MOPS, pH 7.2)Properties: Colorless liquids; Store at 4°C; Preservative-free, use within three months of receipt.