Learn More
TRIBIOSCIENCE INC Calcium Chloride (Sterile, Tissue Culture Grade) 1.25M 100ML
SDP
Catalog No. NC3343209
Encompass
Tribioscience’s CaCl2 Solution at a concentration of 1.25M is used for chemical transfection, including calcium transfection, calcium phosphate transfection, mammalian cell transfection, virus production, lentivirus and retrovirus transfection, and AAV transfection.
Catalog No. NC3343209 Supplier TRIBIOSCIENCE INC Supplier No. TBS5071
Tribioscience’s CaCl2 Solution at a concentration of 1.25M is used for chemical transfection, including calcium transfection, calcium phosphate transfection, mammalian cell transfection, virus production, lentivirus and retrovirus transfection, and AAV transfection.