Learn More
Siemens Healthineers CA Clean™ I Detergent
Catalog No. 23044639 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
23-044-639 50 mL
1 options
Catalog No. 23-044-639 Supplier Siemens Healthineers Supplier No. 10874004
Add to Cart
Add to Cart
For use with Sysmex™ CA Systems
CA Clean™ I detergent is used to cleanse the automated pipets used in Sysmex™ CA systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Detergent
For Use With (Application) Cleansing Automated Pipets
For Use With (Equipment) Sysmex™ CA Systems
Quantity 50 mL
WARNING: Cancer - www.P65Warnings.ca.gov