Learn More
Medchemexpress LLC Ca(II)-EDTA (disodium dihydrate) | 6766-87-6 | 98.0% | 410.30 | C10H12CaN2O8·2Na·2H2O | 5g
SDP
Catalog No. 5000268295
Encompass_Preferred
Ca -EDTA disodium dihydrate (Calcium disodium EDTA dihydrate) is an orally active metal chelating reagent exhibits bactericidal activities against periodontal pathogens Aggregatibacter actinomycetemcomitans Prevotella intermedia and Porphyromonas gingivalis[1] Ca -EDTA disodium dihydrate is effective chelating antidotes for lead-and cadmium poisoning[2][3]
Catalog No. 50-002-68295 Supplier Medchemexpress LLC Supplier No. HYW7498675G
Ca -EDTA disodium dihydrate (Calcium disodium EDTA dihydrate) is an orally active metal chelating reagent exhibits bactericidal activities against periodontal pathogens Aggregatibacter actinomycetemcomitans Prevotella intermedia and Porphyromonas gingivalis[1] Ca -EDTA disodium dihydrate is effective chelating antidotes for lead-and cadmium poisoning[2][3]