Learn More
Hach Company Ca/Mag Indicator Sol 500mL
Supplier:  Hach Company 2241849
Encompass
Calcium/Magnesium Indicator Solution-500 mLFor determination of calcium and magnesium hardness in water by the Calmagite Colorimetric method Hach Method 8030 500 mL bottle
Catalog No. 50-003-04262