Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CA/MAO-B-IN-1 | 3038173-64-4 | 98.3% | 345.37 | C17H15NO5S | 10 MG
SDP

Catalog No. 5000289118
Encompass_Preferred

CA/MAO-B-IN-1 is a research compound that functions as a dual inhibitor of human brain carbonic anhydrases and monoamine oxidase B. It shows low-nanomolar inhibitory potency against several CA isoforms and MAO-B and is used in neuroscience research exploring neuroprotection and amyloid-related toxicity.

  • Dual inhibitory activity against carbonic anhydrases and monoamine oxidase B
  • IC50 values: hCA VB 8.8 nM; MAO-B 7.0 nM; hCA VA 9.6 nM; hCA II 41.1 nM; hCA XII 196 nM
  • Molecular formula C17H15NO5S and molecular weight 345.37
  • High purity (~98% by certificate of analysis)
  • Demonstrated neuroprotective and mitochondrial protective effects in cell models
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month)
  • Supplied as a 10 mg research quantity with solubility/preparation guidance

Catalog No. 50-002-89118 Supplier Medchemexpress LLC Supplier No. HY16338010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CA/MAO-B-IN-1 is a research compound that functions as a dual inhibitor of human brain carbonic anhydrases and monoamine oxidase B. It shows low-nanomolar inhibitory potency against several CA isoforms and MAO-B and is used in neuroscience research exploring neuroprotection and amyloid-related toxicity.

  • Dual inhibitory activity against carbonic anhydrases and monoamine oxidase B
  • IC50 values: hCA VB 8.8 nM; MAO-B 7.0 nM; hCA VA 9.6 nM; hCA II 41.1 nM; hCA XII 196 nM
  • Molecular formula C17H15NO5S and molecular weight 345.37
  • High purity (~98% by certificate of analysis)
  • Demonstrated neuroprotective and mitochondrial protective effects in cell models
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months), -20°C (1 month)
  • Supplied as a 10 mg research quantity with solubility/preparation guidance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.