Learn More
Medchemexpress LLC Ca/mao-b-in-1 | 98.3% | 345.37 | 1ML
SDP
Catalog No. 5000391686
Encompass_Preferred
CA/MAO-B-IN-1 (Compound 78) is a dual inhibitor for human brain carbonic anhydrases (CA) and Monoamine Oxidase-B (MAO-B), with IC50s of 8.8 and 7.0 nM, respectively. This compound demonstrates a human oral absorption of 71.9% based on in silico prediction.
  • Dual inhibitor for human brain carbonic anhydrases (CA) and Monoamine Oxidase-B (MAO-B).
  • IC50s are 8.8 nM for CA and 7.0 nM for MAO-B.
  • Exhibits neuro- and mitochondrial protective effects against Aβ1-42 oligomer-induced neurotoxicity in SH-SY5Y cells.
  • Reverts formation of reactive oxygen species (ROS).
  • In silico prediction suggests 71.9% human oral absorption.
Catalog No. 50-003-91686 Supplier Medchemexpress LLC Supplier No. HY16338010MM/1ML
CA/MAO-B-IN-1 (Compound 78) is a dual inhibitor for human brain carbonic anhydrases (CA) and Monoamine Oxidase-B (MAO-B), with IC50s of 8.8 and 7.0 nM, respectively. This compound demonstrates a human oral absorption of 71.9% based on in silico prediction.
  • Dual inhibitor for human brain carbonic anhydrases (CA) and Monoamine Oxidase-B (MAO-B).
  • IC50s are 8.8 nM for CA and 7.0 nM for MAO-B.
  • Exhibits neuro- and mitochondrial protective effects against Aβ1-42 oligomer-induced neurotoxicity in SH-SY5Y cells.
  • Reverts formation of reactive oxygen species (ROS).
  • In silico prediction suggests 71.9% human oral absorption.