Learn More
Medchemexpress LLC CA/MAO-B-IN-1 | 98.3% | 345.37 | 50 MG
SDP
Catalog No. 5000392995
Encompass_Preferred
CA/MAO-B-IN-1 is a dual inhibitor targeting human brain carbonic anhydrases (CA) and Monoamine Oxidase-B (MAO-B). It demonstrates high potency with IC50s of 8.8 nM and 7.0 nM, respectively, and shows a predicted human oral absorption of 71.9%. This compound is for research use only.
  • Dual inhibitor for human brain carbonic anhydrases and Monoamine Oxidase-B
  • Reverts the formation of reactive oxygen species (ROS)
  • Exhibits neuro- and mitochondrial protective effects
  • Improves cell viability in SH-SY5Y cells damaged by Aβ1-42
Catalog No. 50-003-92995 Supplier Medchemexpress LLC Supplier No. HY16338050MG
CA/MAO-B-IN-1 is a dual inhibitor targeting human brain carbonic anhydrases (CA) and Monoamine Oxidase-B (MAO-B). It demonstrates high potency with IC50s of 8.8 nM and 7.0 nM, respectively, and shows a predicted human oral absorption of 71.9%. This compound is for research use only.
  • Dual inhibitor for human brain carbonic anhydrases and Monoamine Oxidase-B
  • Reverts the formation of reactive oxygen species (ROS)
  • Exhibits neuro- and mitochondrial protective effects
  • Improves cell viability in SH-SY5Y cells damaged by Aβ1-42