Learn More
Lovibond™ CA MG Hardness Set
Catalog No. 01670222 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
10 to 360 mg/L CaCO3
Catalog No. Range
01-670-222 10 to 360 mg/L CaCO3
1 options
This item is not returnable. View return policy
Catalog No. 01-670-222 Supplier Lovibond™ Supplier No. 475100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Unlike other Calmagite test methods, this simplified test method requires only 10 mL of sample and doesn't use additional laboratory equipment for sample preparation. Because the test is carried out in a single vial, glass variation is minimized and a low level of detection is possible.
  • Ideal for pre-treatment of Reverse Osmosis (RO) and water softening systems, this reagent set is used to determine the concentration of calcium and magnesium hardness in a water sample.
  • This easy to use colorimetric test contains 4 liquid reagent solutions for performing up to 100 tests - don't waste time counting drops while waiting for a color change or having to manually calculate the final result.
  • Simplified method for analyzing Calcium and Magnesium Hardness
  • Easy to use colorimetric test
  • Ideal for clean water applications such as pretreatment of RO water and softening systems
Specifications
Analytes Calcium and Magnesium Hardness
For Use With (Application) Analysis of Feedwater for Boiler and Cooling Systems
For Use With (Equipment) Lovibond Spectrophotometers and Colorimeters
No. of Tests 26
Range 10 to 360 mg/L CaCO3
Test Method Calmagite
Type Colorimetric Test