Learn More
Description
- Ideal for analyzing water intended for pre-treatment of Reverse Osmosis (RO) and water softening systems, this reagent set is used to determine the concentration of calcium and magnesium hardness in a water sample.
- Easy dispensing with dropper tip bottle
- Liquid reagent dissolves and mixes easily into sample
- Part of a simplified method for analyzing Calcium and Magnesium Hardness
Specifications
Specifications
| Analytes | Calcium and Magnesium Hardness |
| Description | Solution 1 of 4 in Lovibond's CA MG Hardness Set for measuring Calcium and Magnesium Hardness in a water sample. |
| For Use With (Application) | Pre-Treatment of RO Water and Softening Systems |
| For Use With (Equipment) | Lovibond Spectrophotometers and Colorimeters |
| Quantity | 15 mL |
| Range | 0.05 to 4 mg/L CaCO3 |
| Test Method | Calmagite |
| Type | Ca/Mg Hardness Solution 1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.