Learn More
Perkin Elmer US LLC Calcium (Ca) - Magnesium (Mg) Lumina Hollow Cathode Lamp
Catalog No. 509044274
Encompass_Preferred
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Calcium (Ca) and Magnesium (Mg). Please note that multi-element lamps are not designed to be used in a furnace system. Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.
Catalog No. 50-904-4274 Supplier Perkin Elmer US LLC Supplier No. N3050202
Multi-Element Lumina™ Hollow Cathode Lamp (HCL) for the detection of elemental Calcium (Ca) and Magnesium (Mg). Please note that multi-element lamps are not designed to be used in a furnace system. Every genuine PerkinElmer lamp is designed for use with and tested on our spectrometers to assure compatibility and the highest performance. We offer a wide range of single-element and multi-element HCLs, which are ideal for determining most elements by atomic absorption spectroscopy. To prolong the life of a Lumina™ Hollow Cathode Lamp (HCL), we produce lamps with larger internal volume so that a greater supply of fill gas at optimum pressure is available. The larger the lamp, the greater the inert gas volume - and the longer the lamp lifetime. Note: Lumina™ Hollow Cathode Lamps may also be used with all earlier AA spectrometers that do not have automatic code reading capability by using an appropriate adapter cable.