Learn More
ATCC Ca Ski; Cervical Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRMCRL1550
Encompass
Ca Ski is an epithelial cell that was isolated from the cervix of a 40-year-old, White female with epidermoid. The cell line can be used in testing and calibration in ISO 17025-accredited laboratories, to challenge assay performance, validate or compare test methods, and establish sensitivity, linearity, and specificity during assay validation or implementation.
Catalog No. 50-238-2196