Learn More
MARSHALL SCIENTIFIC Polyscience CA05A1P1-41AA1N Durachill Chiller
SDP
Catalog No. NC2591255
Encompass_Preferred
Stable and reliable cooling for many common heat removal applications, including laser etching, AA furnaces, ICP, rotary evaporators, vacuum systems, reaction vessels, plasma etching, and condenser cooling.
Catalog No. NC2591255 Supplier MARSHALL SCIENTIFIC Supplier No. CA05A1P141AA1N
Add to Cart
Add to Cart
Request a Quote
Stable and reliable cooling for many common heat removal applications, including laser etching, AA furnaces, ICP, rotary evaporators, vacuum systems, reaction vessels, plasma etching, and condenser cooling.