Learn More
Aviva Systems Biology Corp CA1 Antibody - N-terminal region (ARP58432_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58432_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CA1. Alternate names: Car1, CAB, CA-IApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Rat, Sheep
Catalog No. 50-014-62305