Learn More
Medchemexpress LLC CA12/Carbonic Anhydr 10ug
SDP
Catalog No. 5000196590
Encompass_Preferred
Recombinant human carbonic anhydrase 12 (CA12) expressed in Pichia pastoris and bearing an N-terminal 6xHis tag. Supplied lyophilized in a Tris/NaCl/trehalose formulation, the protein is >90% pure (reducing SDS-PAGE), spans residues A25-S301, and has an approximate molecular weight of 33.1 kDa. Intended for biochemical characterization, enzymatic assays, and antibody development.
  • Expressed in Pichia pastoris for eukaryotic folding and post-translational modifications.
  • N-terminal His tag simplifies purification and detection.
  • Lyophilized formulation improves long-term stability and transport.
  • High purity suitable for biochemical assays and antibody production.
  • Recommended storage at -20°C or -80°C after aliquoting.
Catalog No. 50-001-96590 Supplier Medchemexpress LLC Supplier No. HYP7172010UG
Recombinant human carbonic anhydrase 12 (CA12) expressed in Pichia pastoris and bearing an N-terminal 6xHis tag. Supplied lyophilized in a Tris/NaCl/trehalose formulation, the protein is >90% pure (reducing SDS-PAGE), spans residues A25-S301, and has an approximate molecular weight of 33.1 kDa. Intended for biochemical characterization, enzymatic assays, and antibody development.
  • Expressed in Pichia pastoris for eukaryotic folding and post-translational modifications.
  • N-terminal His tag simplifies purification and detection.
  • Lyophilized formulation improves long-term stability and transport.
  • High purity suitable for biochemical assays and antibody production.
  • Recommended storage at -20°C or -80°C after aliquoting.