Learn More
Medchemexpress LLC CA12/Carbonic Anhydr 50ug
SDP
Catalog No. 5000181869
Encompass_Preferred
CA12 (or carbonic anhydrase 12) plays a crucial role in the reversible hydration of carbon dioxide catalyzing its conversion into bicarbonate ions and protons This enzymatic activity is essential for key physiological processes including regulating pH and maintaining acid-base balance CA12/Carbonic Anhydrase 12 Protein Human (HEK293 His) is the recombinant human-derived CA12/Carbonic Anhydrase 12 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81869 Supplier Medchemexpress LLC Supplier No. HYP7286650UG
CA12 (or carbonic anhydrase 12) plays a crucial role in the reversible hydration of carbon dioxide catalyzing its conversion into bicarbonate ions and protons This enzymatic activity is essential for key physiological processes including regulating pH and maintaining acid-base balance CA12/Carbonic Anhydrase 12 Protein Human (HEK293 His) is the recombinant human-derived CA12/Carbonic Anhydrase 12 protein expressed by HEK293 with C-His labeled tag