Learn More
Medchemexpress LLC CA12/Carbonic Anhydr 5ug
SDP
Catalog No. 5000196243
Encompass_Preferred
Recombinant human carbonic anhydrase XII corresponding to residues A25-S301, expressed in Pichia pastoris and supplied as an N-terminal 6xHis-tagged, lyophilized protein. The preparation is glycosylated, ~33.1 kDa, >90% pure by reducing SDS-PAGE, formulated in Tris-HCl/NaCl with trehalose, and has endotoxin <1 EU/μg. Intended for research applications such as biochemical assays, antibody validation, and structural studies.
  • Expressed in Pichia pastoris for eukaryotic glycosylation.
  • Contains an N-terminal 6xHis tag for affinity purification.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • High purity (>90%) suitable for biochemical and immunoassays.
  • Low endotoxin level (<1 EU/μg) for sensitive downstream applications.
  • Stable storage at -20°C; aliquot to minimize freeze-thaw cycles.
Catalog No. 50-001-96243 Supplier Medchemexpress LLC Supplier No. HYP717205UG
Recombinant human carbonic anhydrase XII corresponding to residues A25-S301, expressed in Pichia pastoris and supplied as an N-terminal 6xHis-tagged, lyophilized protein. The preparation is glycosylated, ~33.1 kDa, >90% pure by reducing SDS-PAGE, formulated in Tris-HCl/NaCl with trehalose, and has endotoxin <1 EU/μg. Intended for research applications such as biochemical assays, antibody validation, and structural studies.
  • Expressed in Pichia pastoris for eukaryotic glycosylation.
  • Contains an N-terminal 6xHis tag for affinity purification.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • High purity (>90%) suitable for biochemical and immunoassays.
  • Low endotoxin level (<1 EU/μg) for sensitive downstream applications.
  • Stable storage at -20°C; aliquot to minimize freeze-thaw cycles.