Learn More
Medchemexpress LLC CA125 Human Biotin 20ug | 20ug
SDP
Catalog No. 5000193925
Encompass_Preferred
The CA125 protein acts by binding to MSLN and plays a crucial role in forming a protective and lubricating barrier on mucosal surfaces This interaction promotes heterotypic cell adhesion and is suggested to play an important role in peritoneal metastasis of ovarian cancer CA125 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived CA125 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-93925 Supplier Medchemexpress LLC Supplier No. HYP7817820UG
The CA125 protein acts by binding to MSLN and plays a crucial role in forming a protective and lubricating barrier on mucosal surfaces This interaction promotes heterotypic cell adhesion and is suggested to play an important role in peritoneal metastasis of ovarian cancer CA125 Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived CA125 protein expressed by HEK293 with C-Avi C-His labeled tag