Learn More
Medchemexpress LLC Cancer antigen 125 (MUC16), recombinant human, biotinylated | >95.0% | 50UG
SDP
Catalog No. 5000177144
Encompass_Preferred
Recombinant human CA125 (MUC16) is a biotinylated protein fragment (~685 amino acids) produced in HEK293 cells with C-terminal His and Avi tags. It is intended for binding studies, assay development, and detection workflows that use biotin-streptavidin capture.
  • Biotinylated for streptavidin-based capture and detection.
  • Expressed in HEK293 cells to provide mammalian glycosylation.
  • C-terminal His and Avi tags for purification and site-specific biotinylation.
  • Supplied at laboratory quantity suitable for assay development.
  • High purity as determined by reducing SDS-PAGE (>95%).
  • Molecular weight range 90-140 kDa reflecting glycosylated forms.
Catalog No. 50-001-77144 Supplier Medchemexpress LLC Supplier No. HYP70096050UG
Recombinant human CA125 (MUC16) is a biotinylated protein fragment (~685 amino acids) produced in HEK293 cells with C-terminal His and Avi tags. It is intended for binding studies, assay development, and detection workflows that use biotin-streptavidin capture.
  • Biotinylated for streptavidin-based capture and detection.
  • Expressed in HEK293 cells to provide mammalian glycosylation.
  • C-terminal His and Avi tags for purification and site-specific biotinylation.
  • Supplied at laboratory quantity suitable for assay development.
  • High purity as determined by reducing SDS-PAGE (>95%).
  • Molecular weight range 90-140 kDa reflecting glycosylated forms.