Learn More
Medchemexpress LLC CA125 Human HEK293 5ug | 5ug
SDP
Catalog No. 5000185775
Encompass_Preferred
CA125 is a giant mucin-like glycoprotein expressed by epithelial ovarian neoplasms and cells which is an antigenic tumor marker CA125 provides a protective lubricating barrier against particles and infectious agents at mucosal surfaces CA125 binds to mesothelin (MSLN) mediates heterotypic cell adhesion contributing to the metastasis of ovarian cancer to the peritoneum by initiating cell attachment to the mesothelial epithelium CA125 is widely used for monitoring with ovarian epithelial cancer CA125 Protein Human (HEK293 His) is a recombinant human cancer antigen 125 (CA125) with a His label and is expressed in HEK293 cells[1][2][3][4][5][6][7]
Catalog No. 50-001-85775 Supplier Medchemexpress LLC Supplier No. HYP76965UG
CA125 is a giant mucin-like glycoprotein expressed by epithelial ovarian neoplasms and cells which is an antigenic tumor marker CA125 provides a protective lubricating barrier against particles and infectious agents at mucosal surfaces CA125 binds to mesothelin (MSLN) mediates heterotypic cell adhesion contributing to the metastasis of ovarian cancer to the peritoneum by initiating cell attachment to the mesothelial epithelium CA125 is widely used for monitoring with ovarian epithelial cancer CA125 Protein Human (HEK293 His) is a recombinant human cancer antigen 125 (CA125) with a His label and is expressed in HEK293 cells[1][2][3][4][5][6][7]