Learn More
Biogenex Laboratories CA125 : Ov185:1 : 6mL
SDP
Supplier:  Biogenex Laboratories AM4295M
Encompass_Preferred
Monoclonal antibody Ov185:1 reacts with repetitive protein determinantexpressed in the protein core of the CA125 human ovarian cancer antigen.This marker is usually associated with ovarian epithelial malignancies.Immunohistochemistry with CA125 antibody in conjunction with othermarkers was found to be useful in tracing the origin of adenocarcinomawith unknown primary site. Carcinoembryonic antigen, CA 125,surfactant, E-cadherin, N-cadherin,and vimentin markers are found to beuseful in differentiating epithelial tumors of lung versusovarian origin.The higher rate of positive staining was also seen in endometrial cancers.
Catalog No. 50-197-8855