Learn More
Genscript Corporation CA15-3 AntibodyA5D8 mAb Mo
Catalog No. 5000058318
Encompass
CA15-3 (Carcinoma antigen 15-3) is also known as MUC 1 antigen It is a membrane-anchored mucin-type glycoprotein present in malignant cells It is used as tumor marker to monitor breast cancer treatment and disease recurrence
Catalog No. 50-000-58318 Supplier Genscript Corporation Supplier No. V0860310
CA15-3 (Carcinoma antigen 15-3) is also known as MUC 1 antigen It is a membrane-anchored mucin-type glycoprotein present in malignant cells It is used as tumor marker to monitor breast cancer treatment and disease recurrence