Learn More
Genscript Corporation CA15-3 AntibodyA5D8 mAb Mo
Supplier:  Genscript Corporation V08603100
Encompass
CA15-3 (Carcinoma antigen 15-3) is also known as MUC 1 antigen It is a membrane-anchored mucin-type glycoprotein present in malignant cells It is used as tumor marker to monitor breast cancer treatment and disease recurrence
Catalog No. 50-000-59184
Add to Cart
Request a Quote