Learn More
Biogenex Laboratories CA19-9 : C241:5:1:4 : 6mL
SDP
Supplier:  Biogenex Laboratories AM4245M
Encompass_Preferred
Carcinoma Antigen 19-9 (CA19-9) is a carbohydrate antigen that reactsspecifically with Sialyl Lewis-containing glycolipids and has been isolatedand characterized as the oligosaccharide sialylazed lacto-N-fucopentose IIantigen. This monoclonal antibody is directed against the CA 19-9antigen, which is expressed in pancreatic carcinomas, hepatobillarycarcinomas, the tumor cells of colorectal and gastric. It can also be foundin chronic pancreatitis and in healthy colonic mucosa of patients withcolorectal cancer.
Catalog No. 50-197-8854