Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM125P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 121SLE. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Stomach or Colon Carcinoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Precipitin lines obtained after immuno-diffusion using MAb 116-NS-19-9, mucins isolated from an ovarian cyst of a Lewis A+B- patient (0Le). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C.
Catalog No. 50-188-5408