Learn More
Aviva Systems Biology Corp CA2 Antibody - C-terminal region (ARP60094_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60094_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CA2. Alternate names: CA-II, CAII, Car2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat, Sheep
Catalog No. 50-286-938