Learn More
Medchemexpress LLC Ca2+ channel agonist 1 | 1402821-24-2 | 98.9% | 354.45 | 1 ML
SDP
Catalog No. 5000403688
Encompass_Preferred
Ca2+ channel agonist 1 is an agonist of N-type Ca2+ channel and an inhibitor of Cdk2, with EC50s of 14.23 μM and 3.34 μM, respectively. It is used as a potential treatment for motor nerve terminal dysfunction.
  • Agonist of N-type Ca2+ channel
  • Inhibitor of Cdk2
  • Potential treatment for motor nerve terminal dysfunction
  • Exhibits a ca. 2-fold increased agonism and a 22-fold decreased cdk2 kinase activity versus the standard, (R)-roscovitine
Catalog No. 50-004-03688 Supplier Medchemexpress LLC Supplier No. HY4107610MM/1ML
Ca2+ channel agonist 1 is an agonist of N-type Ca2+ channel and an inhibitor of Cdk2, with EC50s of 14.23 μM and 3.34 μM, respectively. It is used as a potential treatment for motor nerve terminal dysfunction.
  • Agonist of N-type Ca2+ channel
  • Inhibitor of Cdk2
  • Potential treatment for motor nerve terminal dysfunction
  • Exhibits a ca. 2-fold increased agonism and a 22-fold decreased cdk2 kinase activity versus the standard, (R)-roscovitine