Learn More
Santa Cruz Biotechnology CA215 Antibody (RP 215), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC69849
Encompass
CA215 Antibody (RP 215) is a high quality monoclonal CA215 antibody (also designated CA215 antibody) suitable for the detection of the CA215 protein of human origin. CA215 Antibody (RP 215) is available as both the non-conjugated anti-CA215 antibody form, as well as multiple conjugated forms of anti-CA215 antibody, including agarose, HRP, PE, FITC and multiple Alexa Fluor conjugates.
Catalog No. NC1691886