Learn More
Selleck Chemical LLC CA3 (CIL56) 5mg 300802-28-2
SDP
Supplier:  Selleck Chemical LLC S86615MG
Encompass_Preferred
CA3 (CIL56) has potent inhibitory effects on YAP1/Tead transcriptional activity and primarily targets YAP1 high and therapy-resistant esophageal adenocarcinoma cells endowed with CSC properties. CA3(CIL56) induces ferroptosis and iron-dependent reactive oxygen species (ROS). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8027