Learn More
Aviva Systems Biology Corp CA4 Antibody - middle region (ARP41421_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41421_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CA4. Alternate names: CAIV, RP17, Car4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57841