Learn More
Aviva Systems Biology Corp CA5A Antibody - C-terminal region (ARP60096_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60096_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CA5A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CA5A. Alternate names: CA5, CAV, CAVAApplicationWB. Species reactivity: Human
Catalog No. 50-286-940