Learn More
Hach Company CA610 Fluoride Analyzer w/Rgnt
Catalog No. 5000208092 Shop All Hach Company Products
Encompass
CA610 Fluoride Analyzer with ReagentsIncludes CA610 Fluoride analyzer-one-month supply of reagents-installation kit-annual maintenance kit-Hach user manualThe CA610 Fluoride Analyzer delivers accurate real-time fluoride monitoring for municipal water treatment With advanced ion-selective electrode (ISE) technology it provides dependable results and helps maintain water quality compliance with less effort The CA610 Fluoride Analyzer simplifies fluoride analysis making it essential for monitoring water quality With advanced technology it provides precise reliable results to help ensure clean safe water for communities and industries Meeting strict regulatory standards the CA610 supports water safety with efficiency and confidence
Catalog No. 50-002-08092 Supplier Hach Company Supplier No. 5740001
Add to Cart
Add to Cart
Request a Quote
CA610 Fluoride Analyzer with ReagentsIncludes CA610 Fluoride analyzer-one-month supply of reagents-installation kit-annual maintenance kit-Hach user manualThe CA610 Fluoride Analyzer delivers accurate real-time fluoride monitoring for municipal water treatment With advanced ion-selective electrode (ISE) technology it provides dependable results and helps maintain water quality compliance with less effort The CA610 Fluoride Analyzer simplifies fluoride analysis making it essential for monitoring water quality With advanced technology it provides precise reliable results to help ensure clean safe water for communities and industries Meeting strict regulatory standards the CA610 supports water safety with efficiency and confidence