Learn More
Genscript Corporation CA72-4 Antibody D31 mAb Mo
Supplier:  Genscript Corporation V08002100
Encompass
CA72-4 (Cancer antigen 72-4) also known as Tumor associated glycoprotein (TAG) 72 is a mucin-like protein of high molecular weight (200- 400 kD) CA 72-4 has high specificity for diagnosis of benign diseases It is also a helpful marker in the diagnosis of gastric cancer ovarian carcinoma and colorectal carcinoma
Catalog No. 50-000-57588
Add to Cart
Request a Quote