Learn More
Medchemexpress LLC HY-134923 5mg , CA77.1 CAS:2412270-22-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1349235MG
Encompass_Preferred
HY-134923 5mg CA77.1 CAS: 2412270-22-3 CA77.1 is a potent, brain-penetrant and orally active chaperone-mediated autophagy (CMA) activator with favorable pharmacokinetics. CA77.1 is a derivative of AR7 (HY-101106) and can increase the expression of the lysosomal receptor LAMP2A in lysosomes. CA77.1 improves behavior and neuropathology in PS19 mice model and can be used for alzheimer's disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9708