Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 768MSM1XXP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM487. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: Normal kidney or renal cell carcinoma. Cellular Staining: Cell surface, cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Microsomal fraction of human renal cortical tissue homogenate. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 200kDa.Chromosome: 9p13.3. Gene Symbol: CA9. Gene ID: 768. Ensemble: ENSG00000107159. Uniprot: NP_001207.
Catalog No. 50-188-5297