Learn More
Aviva Systems Biology Corp CAB39L Antibody - N-terminal region (ARP60758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAB39L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CAB39L. Alternate names: FLJ12577, MO25-BETA, MO2L, RP11-103J18.3, bA103J18.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-149