Learn More
Cayman Chemical Cab4537-362humnrecomb 20ug
SDP
Catalog No. 502943771
Encompass_Preferred
Source: Recombinant human C-terminal V5- and His-tagged Cab45 expressed in insect cells • Amino acids: 37-362 (full length) • MW: 42.38 kDa
Catalog No. 50-294-3771 Supplier Cayman Chemical Supplier No. 4082220 UG
Source: Recombinant human C-terminal V5- and His-tagged Cab45 expressed in insect cells • Amino acids: 37-362 (full length) • MW: 42.38 kDa