Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cabamiquine succinate | 2444781-71-7 | 99.8% | 580.65 g·mol-1 | C31H37FN4O6 | 1 ML
SDP

Catalog No. 5000227778
Encompass_Preferred

Cabamiquine succinate is the succinate salt of cabamiquine (DDD107498), an orally active antimalarial research compound. Supplied as a 10 mM solution in DMSO in a 1 mL vial, the product is intended for in vitro pharmacology and antimalarial studies. Handle according to the safety data sheet and institutional guidelines.

  • Supplied as a 10 mM solution in DMSO.
  • High purity (99.8%) suitable for research use.
  • Succinate salt form of cabamiquine.
  • Molecular weight 580.65 g·mol-1 and formula C31H37FN4O6.
  • Intended for in vitro antimalarial and pharmacology studies.

Catalog No. 50-002-27778 Supplier Medchemexpress LLC Supplier No. HY117684A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cabamiquine succinate is the succinate salt of cabamiquine (DDD107498), an orally active antimalarial research compound. Supplied as a 10 mM solution in DMSO in a 1 mL vial, the product is intended for in vitro pharmacology and antimalarial studies. Handle according to the safety data sheet and institutional guidelines.

  • Supplied as a 10 mM solution in DMSO.
  • High purity (99.8%) suitable for research use.
  • Succinate salt form of cabamiquine.
  • Molecular weight 580.65 g·mol-1 and formula C31H37FN4O6.
  • Intended for in vitro antimalarial and pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.