Learn More
Medchemexpress LLC HY-15459 100mg , Cabazitaxel XRP6258;RPR-116258A;taxoid XRP6258 CAS:183133-96-2 Purity:98%
SDP
Catalog No. 501493092
Encompass_Preferred
Medchemexpress, HY-15459 100mg Cabazitaxel XRP6258;RPR-116258A;taxoid XRP6258 CAS:183133-96-2 Formula:C45H57NO14 Purity:98% Cabazitaxel is a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3092 Supplier Medchemexpress LLC Supplier No. HY15459100MG
Medchemexpress, HY-15459 100mg Cabazitaxel XRP6258;RPR-116258A;taxoid XRP6258 CAS:183133-96-2 Formula:C45H57NO14 Purity:98% Cabazitaxel is a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.