Learn More
Medchemexpress LLC Cabazitaxel | 183133-96-2 | C45H57NO14 | 25 MG
SDP
Catalog No. 5000375029
Encompass_Preferred
Cabazitaxel is a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity.
  • Purity of 99.98%
  • Microtubule/tubulin inhibitor
  • Potential antineoplastic activity
  • For research use only
Catalog No. 50-003-75029 Supplier Medchemexpress LLC Supplier No. 25MGHY15459
Cabazitaxel is a semi-synthetic derivative of the natural taxoid 10-deacetylbaccatin III with potential antineoplastic activity.
  • Purity of 99.98%
  • Microtubule/tubulin inhibitor
  • Potential antineoplastic activity
  • For research use only